Learn More
3M™ SecureFit™ 3700 Series Protective Eyewear
Catalog No. 19071170 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Amber
Clear
Gray
Indoor/Outdoor, Gray
Temple Color:
Blue
Gray
Green
Lime Green
Orange
Description:
Alternate Fit
Alternate Fit with Browguard
Browguard
Regular Fit
Lens Coating:
Anti-fog
Scratch-resistant
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Temple Color Description Lens Coating
19-071-170 Clear Lime Green Regular Fit -
19-071-160 Clear Blue Regular Fit Anti-fog
19-071-161 Clear Blue Regular Fit Scratch-resistant
19-071-162 Gray Blue Regular Fit Anti-fog
19-071-163 Gray Blue Regular Fit Scratch-resistant
19-071-164 Clear Blue Browguard Anti-fog
19-071-167 Indoor/Outdoor, Gray Blue Regular Fit Anti-fog
19-071-168 Indoor/Outdoor, Gray Blue Regular Fit Scratch-resistant
19-071-171 Amber Green Regular Fit Anti-fog
19-071-165 Clear Blue Browguard Scratch-resistant
19-071-166 Gray Blue Browguard Anti-fog
19-071-169 Clear Lime Green Regular Fit -
19-071-172 Clear Blue Alternate Fit -
19-071-173 Indoor/Outdoor, Gray Blue Browguard Anti-fog
19-071-180 Clear Blue Alternate Fit with Browguard -
19-071-183 Clear Orange Regular Fit -
19-071-188 Clear Orange Regular Fit -
19-071-189 Clear Gray Browguard Anti-fog
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 missing translation for 'options'
This item is not returnable. View return policy
Catalog No. 19-071-170 missing translation for 'mfr' 3M™ missing translation for 'supplierNo' 7100203193
This item is not returnable. View return policy
Get over-the-glass protection with all day wearability
  • The adjustable ratchet temples provides angle adjustment points on the temple to help create a secure and personalized fit
  • Pressure Diffusion Temple (PDT) Technology helps diffuse pressure over the ear for a secure and comfortable fit
  • Soft touch nosepiece to help provide all-day comfort
  • Polycarbonate Lenses absorb 99.9% of UVA and UVB rays
  • Meets the requirements of ANSI Z87.1-2015 Standard as an Impact Rated Spectacle
  • 3M Catalog# SF3701, SF3702, SF3703, SF3707
TRUSTED_SUSTAINABILITY
Specifications
Description Regular Fit
Lens Tint Clear
Product Type Protective eyewear
For Use With (Application) Automotive, Machining, Transportation, Sawing, Riveting
Frame Color Clear
No. per Case 10/Cs.
Model SF3701AS-GRN
Size Universal
Temple Style 3M™ Pressure Diffusion Temple Technology
Temple Color Lime Green
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection 0.999