Learn More
3M™ SecureFit™ 3700 Series Protective Eyewear
Catalog No. 19071189 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Amber
Clear
Gray
Indoor/Outdoor, Gray
Lens Coating:
Anti-fog
Scratch-resistant
Temple Color:
Blue
Gray
Green
Lime Green
Orange
Description:
Alternate Fit
Alternate Fit with Browguard
Browguard
Regular Fit
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Lens Coating Temple Color Description
19-071-189 Clear Anti-fog Gray Browguard
19-071-160 Clear Anti-fog Blue Regular Fit
19-071-161 Clear Scratch-resistant Blue Regular Fit
19-071-162 Gray Anti-fog Blue Regular Fit
19-071-163 Gray Scratch-resistant Blue Regular Fit
19-071-164 Clear Anti-fog Blue Browguard
19-071-167 Indoor/Outdoor, Gray Anti-fog Blue Regular Fit
19-071-168 Indoor/Outdoor, Gray Scratch-resistant Blue Regular Fit
19-071-171 Amber Anti-fog Green Regular Fit
19-071-165 Clear Scratch-resistant Blue Browguard
19-071-166 Gray Anti-fog Blue Browguard
19-071-169 Clear - Lime Green Regular Fit
19-071-170 Clear - Lime Green Regular Fit
19-071-172 Clear - Blue Alternate Fit
19-071-173 Indoor/Outdoor, Gray Anti-fog Blue Browguard
19-071-180 Clear - Blue Alternate Fit with Browguard
19-071-183 Clear - Orange Regular Fit
19-071-188 Clear - Orange Regular Fit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
This item is not returnable. View return policy
Catalog No. 19-071-189 Supplier 3M™ Supplier No. 7100203174
This item is not returnable. View return policy
Get over-the-glass protection with all day wearability
  • The adjustable ratchet temples provides angle adjustment points on the temple to help create a secure and personalized fit
  • Pressure Diffusion Temple (PDT) Technology helps diffuse pressure over the ear for a secure and comfortable fit
  • Soft touch nosepiece to help provide all-day comfort
  • Polycarbonate Lenses absorb 99.9% of UVA and UVB rays
  • Meets the requirements of ANSI Z87.1-2015 Standard as an Impact Rated Spectacle
  • 3M Catalog# SF3701, SF3702, SF3703, SF3707
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Description Browguard
Lens Tint Clear
Product Type Protective eyewear
For Use With (Application) Automotive, Machining, Transportation, Sawing, Riveting
Frame Color Clear
No. per Case 10/Cs.
Model SF3701XSGAF-RED
Size Universal
Temple Style 3M™ Pressure Diffusion Temple Technology
Temple Color Gray
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400