Learn More
3M™ SecureFit™ 400 Series Safety Glasses
Catalog No. 17623766 Shop All 3M Products
Change view
Click to view available options
Frame Color:
Black, Gray
Black, Green
Black, Yellow
Blue and Gray
Hi-Vis Green and Clear
Light Blue, White
Pink, Black
Yellow, Black
Lens Tint:
Amber
Clear
Gray
Indoor/Outdoor
Indoor/Outdoor Mirror
Orange
Lens Coating:
Anti-fog
Scratch-resistant
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint Lens Coating
17-623-766 Black, Yellow Indoor/Outdoor Mirror Scratch-resistant
19-070-944 Yellow, Black Amber Scratch-resistant
19-070-947 Blue and Gray Indoor/Outdoor Scratch-resistant
19-070-948 Blue and Gray Clear Anti-fog
19-070-949 Blue and Gray Clear Scratch-resistant
19-070-951 Black, Green Gray Anti-fog
19-070-952 Black, Green Clear Anti-fog
17-623-767 Black, Yellow Clear Anti-fog
17-623-768 Black, Yellow Gray Anti-fog
19-070-942 Black, Green Gray Scratch-resistant
19-070-943 Pink, Black Gray -
19-070-945 Black, Gray Orange Scratch-resistant
19-070-946 Light Blue, White Clear -
19-070-950 Hi-Vis Green and Clear Clear -
19-070-953 Black, Green Clear -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Catalog No. 17-623-766 Supplier 3M™ Supplier No. 7100112436
Add to Cart
Add to Cart
This item is not returnable. View return policy
3M™ SecureFit™ 400 Series Safety Glasses offer models with stylized colors, proprietary Pressure Diffusion Temple (PDT) technology for comfort, and a contoured brow guard to help provide additional coverage from flying particles.
  • Soft adjustable nosepiece reduces slippage
  • Dual-material temples provide cushioned comfort plus enhanced style
  • Pressure Diffusion Temple Technology diffuses pressure over ear
  • Polycarbonate lenses absorb 99.9% of UVA and UVB
  • Motion-inspired, contoured piping in bold, modern color palette for infusion of style
  • Weighs less than 1 oz.
  • 3M Catalog# SF401AF , 3M Catalog# SF402AF , 3M Catalog# SF410AS
Applications:
Construction, Manufacturing, Oil & Gas, Transportation.
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant
Lens Tint Indoor/Outdoor Mirror
Frame Color Black, Yellow
Model SF410AS
Lens Material Polycarbonate