Learn More
3M™ SecureFit™ 400 Series Safety Glasses
3M™ SecureFit™ 400 Series Safety Glasses offer models with stylized colors, proprietary Pressure Diffusion Temple (PDT) technology for comfort, and a contoured brow guard to help provide additional coverage from flying particles.
Supplier:  3M™ 7100180051
 View more versions of this product
Catalog No. 19-070-947
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Contoured brow guard rests comfortably against forehead and helps provide additional coverage from flying particles
  • Features proprietary 3M™ Pressure Diffusion Temple Technology which helps diffuse pressure over the ear for a secure, comfortable fitContoured brow guard rests comfortably against forehead and helps provide additional coverage from flying particles
  • Enhanced temple comfort without compromising the security of fit across diverse workforcesContoured brow guard rests comfortably against forehead and helps provide additional coverage from flying particles
  • Soft nosepiece to help reduce slippageContoured brow guard rests comfortably against forehead and helps provide additional coverage from flying particles
  • Applied on both sides of the lens, 3M™ Scotchgard™ Anti-fog is a long-lasting coating that provides better fog and scratch resistance when compared to traditional anti-fog coatings
  • 3M Catalog# SF401XAF-GRN, SF401XAS-GRN, SF401XSGAF-BLU, SF401XSGAF-GRN, SF401XSGAF-WHT, SF402XAF-GRN, SF402XAS-GRN, SF402XSGAF-BLU, SF403XSGAF-YEL, SF407XSGAF-BLU, SF409XAS-PNK, SF416XAS-BLK
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Scratch-resistant
Indoor/Outdoor
Blue and Gray
20/Pk.
Polycarbonate
Yes
ANSI Z87.1-2015
Protective Eyewear
SF407XSGAF-BLU
Pressure Diffusion Temple Technology
Polycarbonate
Frameless
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.