Learn More
3M™ SecureFit™ 600 Series Protective Eyewear with +1.5 Diopter
Catalog No. 17000700 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
17-000-700 Gray
17-000-697 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-000-700 Supplier 3M™ Supplier No. 7100132362
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
3M™ SecureFit™ 600 Series Protective Eyewear with +1.5 Diopter features proprietary 3M™ Pressure Diffusion Temple Technology that helps diffuse pressure over the ear to enhance frame comfort while not compromising the security of fit across a diverse workforce.
  • Features proprietary 3M™ Pressure Diffusion Temple Technology which helps diffuse pressure over the ear for a secure, comfortable fit
  • Scotchgard™ Anti-fog Coating lasts longer than traditional anti-fog coatings, helping to increase eyewear performance in wet and steamy environments
  • The proprietary coating retains its effectiveness for at least 25 washings, allowing workers to wear their eyewear longer
  • Workers can feel confident wearing Scotchgard™ Anti-Fog coated eyewear because it meets the requirements of ANSI Z87.1.2015
  • +1.5 diopter combining eye protection with reading enhancement
  • Meets the requirements of ANSI Z87.1-2015 Standard as an Impact Rated spectacle
  • Grey polycarbonate lens absorbs 99.9% UVA & UVB rays
  • 3M Catalog# SF615GSGAF , 3M Catalog# SF615SGAF
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, UV-treated
Lens Type Magnifying
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Gray
Product Type Safety Glasses
Frame Color Gray
Magnification Power +1.50
Model SF615GSGAF
No. per Pack 20/Pk.
Temple Style SecureFit™
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Half Frame