Learn More
3M™ SecureFit™ Protective Eyewear
Catalog No. 17623701 Shop All 3M Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Scratch-resistant
Lens Tint:
Amber
Clear
Gray
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating Lens Tint
17-623-701 Anti-fog Gray
17-623-700 Anti-fog Clear
17-623-703 Scratch-resistant Clear
17-623-702 Anti-fog Amber
17-623-704 Scratch-resistant Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-623-701 Supplier 3M™ Supplier No. 7100022210
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Pressure Diffusion Temple Technology that helps diffuse pressure over the ear to enhance frame comfort
  • Need to stock multiple frame sizes at work sites eliminated by self-adjusting feature
  • Secure, snug fit prevents slippage even during motion
  • Anti-fog coating ensures moisture free lenses in hot and humid conditions
  • Anti-scratch coating protects lenses from scratches, nicks and scrapes
  • Polycarbonate lenses absorb 99.9% UVA and UVB up to 380nm
  • 3M Catalog# SF201AF, SF201AS, SF202AF, SF202AS, SF203AF
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Certifications/Compliance ANSI Z87.1-2015, CSA Z94.3
Lens Tint Gray
Model SF202AF
Temple Style Proprietary self-adjusting 3M Pressure Diffusion Temple Technology
Frame Type Frameless