Learn More
3M™ SecureFit™ Protective Eyewear
Catalog No. 17623703 Shop All 3M Products
Change view
Click to view available options
Lens Coating:
Anti-fog
Scratch-resistant
Lens Tint:
Amber
Clear
Gray
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating Lens Tint
17-623-703 Scratch-resistant Clear
17-623-700 Anti-fog Clear
17-623-701 Anti-fog Gray
17-623-702 Anti-fog Amber
17-623-704 Scratch-resistant Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-623-703 Supplier 3M™ Supplier No. 7000128262
This item is not returnable. View return policy
Pressure Diffusion Temple Technology that helps diffuse pressure over the ear to enhance frame comfort
  • Need to stock multiple frame sizes at work sites eliminated by self-adjusting feature
  • Secure, snug fit prevents slippage even during motion
  • Anti-fog coating ensures moisture free lenses in hot and humid conditions
  • Anti-scratch coating protects lenses from scratches, nicks and scrapes
  • Polycarbonate lenses absorb 99.9% UVA and UVB up to 380nm
  • 3M Catalog# SF201AF, SF201AS, SF202AF, SF202AS, SF203AF
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant
Certifications/Compliance ANSI Z87.1-2015, CSA Z94.3
Lens Tint Clear
Model SF201AS
No. per Pack 20/Pk.
Temple Style Proprietary self-adjusting 3M Pressure Diffusion Temple Technology
Lens Material Polycarbonate
Frame Type Frameless