Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ SecureFit™ Safety Helmet

Catalog No. 19071072 Shop All 3M Products
Change view
Click to view available options
Color:
Black
Blue
Green
Hi-vis Green
Orange
Red
White
Yellow
Ventilated:
Not Ventilated
Ventilated
Quantity:
10/Cs.
4/Cs.
32 product options available for selection
Product selection table with 32 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Ventilated Quantity
19-071-072 Black Ventilated 4/Cs.
19-071-058 White Not Ventilated 4/Cs.
19-071-066 White Ventilated 4/Cs.
19-071-074 White Not Ventilated 10/Cs.
19-071-082 White Ventilated 10/Cs.
19-071-064 Black Not Ventilated 4/Cs.
19-071-080 Black Not Ventilated 10/Cs.
19-071-088 Black Ventilated 10/Cs.
19-071-060 Blue Not Ventilated 4/Cs.
19-071-068 Blue Ventilated 4/Cs.
19-071-076 Blue Not Ventilated 10/Cs.
19-071-084 Blue Ventilated 10/Cs.
19-071-065 Hi-vis Green Not Ventilated 4/Cs.
19-071-073 Hi-vis Green Ventilated 4/Cs.
19-071-081 Hi-vis Green Not Ventilated 10/Cs.
19-071-089 Hi-vis Green Ventilated 10/Cs.
19-071-063 Orange Not Ventilated 4/Cs.
19-071-071 Orange Ventilated 4/Cs.
19-071-079 Orange Not Ventilated 10/Cs.
19-071-087 Orange Ventilated 10/Cs.
19-071-078 Red Not Ventilated 10/Cs.
19-071-061 Green Not Ventilated 4/Cs.
19-071-069 Green Ventilated 4/Cs.
19-071-077 Green Not Ventilated 10/Cs.
19-071-085 Green Ventilated 10/Cs.
19-071-059 Yellow Not Ventilated 4/Cs.
19-071-067 Yellow Ventilated 4/Cs.
19-071-075 Yellow Not Ventilated 10/Cs.
19-071-120 Yellow Ventilated 10/Cs.
19-071-062 Red Not Ventilated 4/Cs.
19-071-070 Red Ventilated 4/Cs.
19-071-086 Red Ventilated 10/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 32 options available.
32 options
Catalog No. 19-071-072 Supplier 3M™ Supplier No. 7100175568
Add to Cart
Add to Cart

Suspension system incorporates exclusive patented Pressure Diffusion Technology.

  • Works with a variety of 3M faceshields, earmuffs, and other accessories.
  • Meets ANSI Z89.1.
  • Climbing helmet-inspired design.
  • Customize your fit with multiple levels of vertical adjustment.
  • Optional vents on sides and back are designed to allow airflow through the hard hat
  • 3M Catalog# X5001, X5002, X5003, X5004, X5005, X5007, X5012, X5014

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI/ISEA Z89.1-2014 Type 1
Color Black
For Use With (Application) Forestry; Construction; Oil and gas
Material ABS
Model X5012VX-ANSI
Product Type Safety Helmet
Quantity 4/Cs.
Size One Size Fits Most
Style Climbing helmet-inspired design
Suspension Type Pressure Diffusion 6 Point Ratchet
Ventilated Ventilated
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.