Learn More
Phenomenex™ SecurityGuard™ PREP Reverse Phase HPLC Cartridges, Core-Shell C18-Peptide
Shop All Phenomenex Products
Description
- SecurityGuard™ PREP Cartridges for use with Reverse Phase HPLC columns
- All columns are 15 mm in length
Specifications
Specifications
| Quantity | 1 Ea. |
| Particle Size | Proprietary |
| Length (Metric) | 15 mm |
| Type | Guard Cartridge |
| Diameter (Metric) Inner | 21.2 mm |
| Technique | Reverse Phase HPLC |
| Column Type | Guard Column |
| Sorbent | Core-Shell C18-Peptide |
| Product Line | SecurityGuard |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.