Learn More
Phenomenex™ SecurityGuard™ SemiPrep Reverse Phase HPLC Cartridges
Catalog No. 502401056 Shop All Phenomenex Products
Encompass
Change view
Click to view available options
Stationary Phase:
AQ C18
C12
C5
Clarity Oligo-RP
Fusion-RP
Gemini C18
Gemini C6-Phenyl
Gemini NX-C18
PFP
PS C18
Phenyl
Polar-RP
RP-1
Widepore C18
Widepore C4
Widepore C5
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stationary Phase
50-240-1056 Gemini C18
50-239-7478 Widepore C18
50-239-9142 Widepore C4
50-239-9381 C12
50-239-9732 Polar-RP
50-239-7482 Phenyl
50-240-1031 RP-1
50-239-7977 Widepore C5
50-239-8002 C5
50-240-1054 AQ C18
50-239-8920 Fusion-RP
50-240-0042 Clarity Oligo-RP
50-239-9686 Gemini NX-C18
50-239-8262 PFP
50-239-9330 Gemini C6-Phenyl
50-239-8810 PS C18
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 50-240-1056 Supplier Phenomenex™ Supplier No. AJ07598
SecurityGuard™ SemiPrep Cartridges for use with Reverse Phase HPLC columns.
  • SecurityGuard™ SemiPrep Cartridges for use with Reverse Phase HPLC columns
  • Available with multiple stationary phases
  • All columns are 10 x 10 mm
TRUSTED_SUSTAINABILITY
Specifications
Quantity 3/Pk
Particle Size Proprietary
Length (Metric) 10 mm
Type Guard Cartridge
Diameter (Metric) Inner 10 mm
For Use With (Equipment) Standard Gemini C18
Stationary Phase Gemini C18
For Use With (Application) Column protection
Technique Reverse Phase HPLC
Column Type Guard Column
Product Line SecurityGuard