Learn More
Phenomenex™ SecurityGuard™ SemiPrep Reverse Phase HPLC Cartridges, Core-Shell
Catalog No. 502400380 Shop All Phenomenex Products
Encompass
Change view
Click to view available options
Stationary Phase:
Core-Shell Biphenyl
Core-Shell C18
Core-Shell C18-Peptide
Core-Shell EVO C18
Core-Shell F5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stationary Phase
50-240-0380 Core-Shell C18-Peptide
50-239-9625 Core-Shell C18
50-239-9329 Core-Shell Biphenyl
50-239-7127 Core-Shell EVO C18
50-239-8410 Core-Shell F5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 50-240-0380 Supplier Phenomenex™ Supplier No. AJ09317
SecurityGuard™ SemiPrep Cartridges with a core-shell for use with Reverse Phase HPLC columns.
  • SecurityGuard™ SemiPrep Cartridges for use with Reverse Phase HPLC columns
  • Available with multiple stationary phases
  • All columns are 10 x 10 mm
TRUSTED_SUSTAINABILITY
Specifications
Quantity 3/Pk
Particle Size Proprietary
Length (Metric) 10 mm
Type Guard Cartridge
Diameter (Metric) Inner 10 mm
Stationary Phase Core-Shell C18-Peptide
Technique Reverse Phase HPLC
Column Type Guard Column
Product Line SecurityGuard