Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polymedco™ SED-CHEK 2™ ESR Controls
GSA_VASDP

Catalog No. 2246917 Shop All Polymedco Products
Change view
Click to view available options
Type:
Abnormal Control
Bi-Level Control
Normal Control
For Use With (Equipment):
For Sedimat and Manual Procedures
For use with Sedimat™ 15 automated ESR analyzer
Sedimat™ 15 automated ESR analyzer
Quantity:
3 x 8 mL
6 x 8 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment) Quantity
22-469-17 Bi-Level Control Sedimat™ 15 automated ESR analyzer 3 x 8 mL
22-046912 Abnormal Control For Sedimat and Manual Procedures 6 x 8 mL
22-046913 Normal Control For Sedimat and Manual Procedures 6 x 8 mL
22-046911 Bi-Level Control For Sedimat and Manual Procedures 6 x 8 mL
22-469-18 Abnormal Control For use with Sedimat™ 15 automated ESR analyzer 6 x 8 mL
22-469-19 Normal Control For use with Sedimat™ 15 automated ESR analyzer 6 x 8 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22-469-17 Supplier Polymedco™ Supplier No. ESR5CT
Add to Cart
Add to Cart

Unique cell-stabilizing process mimics the rouleaux formation of patient samples

  • Bi-level, whole-blood control designed to monitor ESR procedures
  • Monitor technique as well as environmental and physical factors that adversely affect ESR results
  • Run like a patient sample—not necessary to void diluent from pre-filled tubes
  • Universal control assayed for both automated and manual ESR methods

Order Info

Controls packaged in 6 x 8mL vials

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Cap Type Automatic self-zeroing
Compatible Methods Both automated and manual ESR methods
Content And Storage 20°C to 22°C
For Use With (Equipment) Sedimat™ 15 automated ESR analyzer
Type Bi-Level Control
Intended Use Whole Blood reference control material designed to monitor ESR procedures
Packaging Vial
Quantity 3 x 8 mL
Stability 31 Days Open Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.