Learn More
Description
- Bi-level, whole-blood control designed to monitor ESR procedures
- Monitor technique as well as environmental and physical factors that adversely affect ESR results
- Run like a patient sample—not necessary to void diluent from pre-filled tubes
- Universal control assayed for both automated and manual ESR methods
Order Info
Controls packaged in 6 x 8mL vials
Specifications
Specifications
| Cap Type | Automatic self-zeroing |
| Compatible Methods | Both automated and manual ESR methods |
| Content And Storage | 20°C to 22°C |
| For Use With (Equipment) | For use with Sedimat™ 15 automated ESR analyzer |
| Type | Abnormal Control |
| Intended Use | Whole Blood reference control material designed to monitor ESR procedures |
| Packaging | Vial |
| Quantity | 6 x 8 mL |
| Stability | 31 Days Open Vial |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
