Learn More
Percival Scientific Seed Germination Incubator, GR36L, 29.7 cu. ft., vertical IncuWhite LED lighting, 15 slideable shelves on stainless steel rail assemblies, 5-44° C ±0.5°, IntellusUltra controller
SDP
Supplier:  Percival Scientific GR36L
Encompass_Preferred
The GR36L incubator is designed for seed germination and development. Vertical IncuWhite LEDs are dimmable between 5-100% in 1% increments output with up to 140 µmoles/m2/s irradiance @ 6” from the LEDs. Interior / exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. Fifteen white epoxy coated steel wire shelves slide in and out on stainless steel rail assemblies. Horizontal airflow through rear wall perforated plenum. IntellusUltra control system. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; this also prolongs life of compressor, and eliminates risk of ice build up in coil. Top mounted refrigeration system allows easy access for maintenance and cleaning. For additional options contact sales@percival-scientific.com for a free consultation.
Catalog No. NC2734544
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.