Learn More
Labconco Seismic Kit for Hydraulic Base Stands
GSA_VASDP
Catalog No. 17200228 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Enclosures
Catalog No. For Use With (Equipment)
17-200-228 Enclosures
1 options
Catalog No. 17-200-228 Supplier Labconco Supplier No. 5190300
Add to Cart
Add to Cart
Seismic Kit for Hydraulic Base Stands secures hydraulic base stands to the floor. Includes 4 stainless steel brackets and hardware for attachment to stand. Floor hardware and installation required.
  • Seismic Kit for Labconco Hydraulic Base Stands
  • Secures Labconco hydraulic base stand legs to the floor
  • Includes 4 stainless steel brackets and hardware for attachment to stand
  • Floor hardware and installation required
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
For Use With (Equipment) Enclosures
Product Type Seismic Kit