Learn More
TARGETMOL CHEMICALS INC SEL120-34A 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T10744
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SEL120-34A is a selective and ATP-competitive inhibitor of CDK8 with IC50s of 4.4 nM 10.4 nM and 1070 nM for CDK8/CycC CDK19/CycC and CDK9/cycT. SEL120-34A possesses antitumor activity. purity: 99%
Catalog No. 50-258-1676
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.