Learn More
Bachem (D-Pro7)-Angiotensin I/II (1-7) Trifluoroacetate
SDP
Supplier:  Bachem 4053534.0001
Encompass_Preferred
(D-Pro7)-Angiotensin I/II (1-7) Trifluoroacetate, H-Asp-Arg-Val-Tyr-Ile-His-D-Pro-OH, 586962-44-9, C41H62N12O11, Mr 899.02, 1mg. DPro7-angiontensin-(1-7). Selective angiotensin (1-7) antagonist.
Catalog No. 50-194-6749
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.