Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selectophore™ 2-Nitrophenyl octyl ether, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111014085 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4085 100 mL
11-101-4087 5 mL
11-101-4086 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-4085 Supplier MilliporeSigma Supelco Supplier No. 73732100ML
Add to Cart
Add to Cart

Chemical Identifiers

MDL Number MFCD00014693
Synonym 1-Nitro-2-octyloxybenzene

Specifications

Density 1.04 g/mL (at 25°C)
CAS 37682-29-4
Boiling Point 197°C to 198°C (11 mmHg)
Flash Point 113°C - Closed Cup
Linear Formula O2 NC6H4 O(CH2)7 CH3
Quantity 100 mL
Formula Weight 251.32
Percent Purity ≥99.0% (GC); ≥99.0%
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.