Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selectophore™ Dichloromethane, 99.5%, MilliporeSigma™ Supelco™

Catalog No. 111018930 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8930 500 mL
11-101-9812 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 11-101-8930 Supplier MilliporeSigma Supelco Supplier No. 66742500ML
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Description Solvent for preparation of membranes
CAS 75-09-2
Product Line Selectophore
Ignition Point 1223°F
Solution Contains ˜0.005% amylene as stabilizer
Vapor Pressure 24.45 psi (55°C)
Vapor Density 2.9 (vs. air)
Density 1.324-1.326 g/mL (at 20°C)
Linear Formula CH2 Cl2
MDL Number MFCD00000881
Melting Point -97°C
Boiling Point 39.8°C to 40°C
For Use With (Equipment) Ion Selective Electrode
Synonym Methylene chloride
Type Organic Standard
Quantity 500 mL
Formula Weight 84.93
Percent Purity ≥99.5% (GC)
Show More Show Less

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.