Learn More
Selectophore™ Dipentyl phthalate, 99.0%, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Density | 1.025 g/mL (at 20°C) |
| Linear Formula | C18H26O4 |
| Refractive Index | n20/D 1.490 |
| Quantity | 5 mL |
| Formula Weight | 306.40 |
| Percent Purity | ≥99.0% (GC) |
| Grade | Selectophore |
| Product Line | Selectophore |
Safety and Handling
| RTECSNumber | TI1930000 |
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.