Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selectophore™ Silanization solution I ∼5 (dimethyldichlorosilane in heptane), MilliporeSigma™ Supelco™

Catalog No. 111018310 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8310 1 L
11-101-9366 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8310 Supplier MilliporeSigma Supelco Supplier No. 851261L
Add to Cart
Add to Cart

Visit our Sensor Applications portal to learn more.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00000491
Synonym Dichlorodimethylsilane solution; Dimethyldichlorosilane solution

Specifications

CAS 75-78-5
Quantity 1 L
Linear Formula C2H6 Cl2 Si
Flash Point -4°C
Formula Weight 129.06
Percent Purity 5% (dimethyldichlorosilane in heptane)
Grade Selectophore
Product Line Selectophore

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.