Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selegiline Hydrochloride, USP, 98-101%, Spectrum™ Chemical
SDP

Catalog No. 18606167 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
1 kg
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-167 1 g Amber Glass Bottle
18-606-169 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-606-167 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S15981GM

S1598, 14611-52-0, C13H17N·HCl

Spectrum™ Chemical Selegiline Hydrochloride, USP is an enzyme blocker used in treating depression, dementia, and Parkinson’s disease. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 14611-52-0
Molecular Formula C13H18ClN
Molecular Weight (g/mol) 223.74
MDL Number MFCD00069299
InChI Key IYETZZCWLLUHIJ-UTONKHPSSA-N
IUPAC Name hydrogen methyl[(2R)-1-phenylpropan-2-yl](prop-2-yn-1-yl)amine chloride
SMILES [H+].[Cl-].C[C@H](CC1=CC=CC=C1)N(C)CC#C

Specifications

CAS Max % 1
Ignition Residue 0.002
Loss on Drying 0.01
Packaging Amber Glass Bottle
Quantity 1 g
Specific Rotation -10.0° to -12°
Percent Purity 98 to 101%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.