Learn More
Specifications
Specifications
| Physical Form | Solid |
| Quantity | 250 g |
| Formula Weight | 78.96 |
| Odor | Odorless |
| Packaging | Plastic bottle |
| Appearance | Dark gray solid |
| Chemical Name or Material | Selenium |
Safety and Handling
| Hazard Category | H331: Toxic if inhaled. H301: Toxic if swallowed. H373: May cause damage to organs through prolonged or repeated exposure. H413: May cause long lasting harmful effects to aquatic life. |
| EINECSNumber | 231-957-4 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.