Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selenium Standard, 1mL = 1mg Se, 1000ppm Se, Ricca Chemical
SDP

Catalog No. p-3252248 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Poly natural
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6700-4 120 mL Poly natural
6700-16 500 mL Poly natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6700-4 Supplier Ricca Chemical Company Supplier No. 67004

Chemical Identifiers

CAS 7446-08-4 , 7732-18-5
Molecular Formula O2Se
Molecular Weight (g/mol) 110.97
MDL Number MFCD00003562
InChI Key JPJALAQPGMAKDF-UHFFFAOYSA-N
PubChem CID 24007
IUPAC Name (oxo-λ⁴-selanylidene)oxidane
SMILES O=[Se]=O

Specifications

Packaging Poly natural
Concentration or Composition Notes 1,000ppm Se
Equivalence 1 mL = 1 mg Se
Physical Form Liquid
Color Colorless
Quantity 120 mL
Chemical Name or Material Selenium Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.