Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Selenium Tetrachloride, MP Biomedicals

Catalog No. icn217661 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN217661 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN217661 Supplier MP Biomedicals, Inc Supplier No. 0521766110

Chemical Identifiers

CAS 10026-03-6
Molecular Formula Cl4Se
Molecular Weight (g/mol) 220.771
InChI Key LNBXMNQCXXEHFT-UHFFFAOYSA-N
Synonym selenium tetrachloride, selenium chloride secl4 , t-4, unii-4gb8868p5j, selenium iv chloride 1:4, secl4, tetrachloro-, e4-selane, selenium iv chloride, selenium iv chloride-8 mesh, selenium iv chloride trace metals basis 25g
PubChem CID 66205
SMILES Cl[Se](Cl)(Cl)Cl

Specifications

Quantity 10 g
Formula Weight 220.77
Chemical Name or Material Selenium Tetrachloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.