Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Selenocystine | 2897-21-4 | MFCD00055925 | 99.0% | 334.09 g/mol | C6H12N2O4Se2 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1130645MG

Encompass_Preferred

Selenocystine is the oxidized dimer of selenocysteine, a selenium-containing amino acid used as a biochemical research reagent and investigational anticancer agent. It is employed in studies of selenium biology, redox chemistry, and cellular responses to oxidative stress (CAS 2897-21-4).

  • High purity: 99.0%.
  • Accurate molecular weight: 334.09 g/mol.
  • Chemical formula: C6H12N2O4Se2.
  • Suitable for cellular and biochemical assays investigating redox and DNA-damage pathways.
  • Available in small research pack sizes such as 5 mg.

Catalog No. 50-002-62999


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.