Learn More
Description
- Dry erase panels for displaying metrics and schedules that are frequently updated
- For shop floor improvement ideas and maintenance requests
- Adhesive backing allows for easy application for use on Lean Communication Boards
Specifications
Specifications
| Color | White |
| For Use With (Application) | Displaying frequently changed metrics and schedules or for shop floor improvement ideas and maintenance requests |
| For Use With (Equipment) | Lean Communication Boards |
| Quantity | 5/Pk. |
| Dimensions (L x W) | 11 x 9 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.