Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Self-Standing Graduated Centrifuge Tubes with Screw Cap

Catalog No. 220101225
Change view
Click to view available options
Closure Type:
Attached Screw Cap
Screw Cap
Closure Color:
Natural
Red
Sterility:
Non-sterile
Sterile
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type Closure Color Sterility
22-010-1225 Screw Cap Natural Non-sterile
22-170-199 Screw Cap Red Non-sterile
22-170-261 Attached Screw Cap Red Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-010-1225 Supplier Globe Scientific Supplier No. 6255MF
Add to Cart
Edge
Add to Cart

Leak-resistant and ideal for centrifugation, transport, storage and collection of specimen

  • Material: Polypropylene
  • Cap is made from High Density Polyethylene (HDPE)
  • Autoclavable (without screw cap)
  • Sterile tubes are RNase/DNase and Pyrogen Free
  • Spin rate: 5000rpm (6,160 x g)
  • Conical bottom with skirt
  • Dimensions: 30 x 115mm

Specifications

Closure Color Natural
Capacity (Metric) 50 mL
Material Polypropylene
No. per Case 500
Sterility Non-sterile
Closure Type Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.