Learn More
Globe Scientific Self-Standing Transport Tubes with Printed Graduations
Catalog No. 21403109
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
21-403-109 5 mL
21-403-110 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-403-109 Supplier Globe Scientific Supplier No. 6104SP
Only null left
Add to Cart
Add to Cart
These transparent polypropylene (PP) tubes allow unobstructed viewing of the sample with clearly printed graduations
  • These disposable screwcap microcentrifuge tubes are available sterile or non-sterile
  • The sterile tubes include a cap with nitrile o-ring
  • The non-sterile tubes and caps are sold separately so that you have a choice of cap styles (with o-ring or without)
  • The vials are shatter-proof and may be exposed to freezing temperatures
  • They feature clearly printed graduations and white writing surfaces
Specifications
Autoclavable Not Autoclavable
Capacity (Metric) 5 mL
Bottom Shape Conical (Self-standing)
Material Polypropylene
Thread Style External
Sterility Sterile
Graduated Yes
Product Type Transport Tube (Self-Standing)