Learn More
Globe Scientific Self-Standing Transport Tubes with Printed Graduations
Shop All Globe Scientific ProductsDescription
- These disposable screwcap microcentrifuge tubes are available sterile or non-sterile
- The sterile tubes include a cap with nitrile o-ring
- The non-sterile tubes and caps are sold separately so that you have a choice of cap styles (with o-ring or without)
- The vials are shatter-proof and may be exposed to freezing temperatures
- They feature clearly printed graduations and white writing surfaces
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Capacity (Metric) | 10 mL |
| Bottom Shape | Conical (Self-standing) |
| Material | Polypropylene |
| Thread Style | External |
| Sterility | Sterile |
| Graduated | Yes |
| Product Type | Transport Tube (Self-Standing) |