Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Sell (Rat) ELISA Kit

Catalog No. 89028564 Shop All Abnova Corporation Products
Change view
Click to view available options
Detection Limit:
<20 pg/ml
6 pg/mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detection Limit
89-028-564 6 pg/mL
89-101-096 <20 pg/ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-028-564 Supplier Abnova™ Supplier No. KA1830

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

In-Vitro Enzyme-Linked Immunosorbent assay for the quantitative measurement of Rat Sell

Specifications

Content And Storage Store the kit at -20°C. Standard should be stored at -20°C or -80°C. Avoid repeated freezing and thawing.
Description Sell (Rat) ELISA Kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of rat Sell.
Conjugate Biotin
Cross Reactivity Rat
Detection Method Colorimetric
Product Type Sell (Rat) ELISA Kit
Sample Type Cell Culture Supernatant, Plasma, Serum
Sufficient For 96
For Use With (Application) Quantification
Kit Contents 96-well Antibody coated microwell, 25mL Wash Buffer 20X, 2 vials standard, 30mL assay diluent A, 15mL assay diluent B, 2 vials detection Antibody MSP, 8μL HRP Streptavidian concentrate, 12mL TMB one step Substrate reagent, 8mL stop solut
Quantity 96 Assays
Regulatory Status For research use only (RUO)
Sample Volume 100 μL
Detection Limit 6 pg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.