Learn More
Specifications
Specifications
| Calibration Range | 625 to 40000 pg/mL |
| Content And Storage | Store the kit at -20°C |
| Description | Sell (Rat) is a sandwich enzyme immunoassay for the quantitative measurement of rat Sell. |
| Conjugate | Biotin |
| Cross Reactivity | Rat |
| Detection Method | Colorimetric |
| Product Type | SELL (Rat) ELISA kit |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum |
| Sufficient For | 96 |
| For Use With (Application) | Quantification |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.