Learn More
TARGETMOL CHEMICALS INC SELUMETINIB 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6218
Encompass_Preferred
Also available in 25 mg 50 mg 100 mg 500 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Selumetinib (AZD6244) (AZD6244) is an effective specific inhibitor of MEK1 and ERK1/2 phosphorylation (IC50 14/10 nM). purity: 99%
Catalog No. 50-257-6572
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.