Learn More
Invitrogen™ SEMA4B Polyclonal Antibody

Description
SEMA4B Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot
Specifications
Specifications
| Antigen | SEMA4B |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 3.185 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | SEMA4B |
| Gene Accession No. | Q9NPR2 |
| Gene Alias | KIAA1745; mKIAA1745; sema C; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4B; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4B; Sema4b; SEMAC; semaphorin 4B; semaphorin-4B; Semaphorin-C; SemC; UNQ749/PRO1480 |
| Gene Symbols | SEMA4B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.