Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEMA4D Monoclonal Antibody (OTI8G5), TrueMAB™, OriGene
SDP

Catalog No. 501683054
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3054 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3054 Supplier OriGene Supplier No. TA809851
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SEMA4D (CD100) is a cell surface receptor for PLXN1B and PLXNB2. It plays an important role in cell-cell signaling and promotes reorganization of the actin cytoskeleton. CD100 also plays a role in axonal growth cone guidance in the developing central nervous system, as well as, regulates dendrite and axon branching and morphogenesis. CD100 is also active in the immune system - it induces B-cells to aggregate and improves thru viability.

Specifications

Antigen SEMA4D
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI8G5
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SEMA4D
Gene Accession No. Q92854
Gene Alias C9orf164, CD100, COLL4, M-sema-G, SEMAJ, coll-4
Gene Symbols SEMA4D
Host Species Mouse
Immunogen Full length human recombit protein of human SEMA4D produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10507
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.