Learn More
Carbosynth LLC. Semaglutide, 1mg, Bolar Exemption applies.
Supplier: Carbosynth LLC. GLP3875PI1MG
Synonyms: H-His-Aib-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Val-Ser-Ser-Tyr-Leu-Glu-Gly-Gln-Ala-Ala-Lys[(AEEA)2-Ï’-Glu-N-(17-carboxy-1-oxoheptadecyl)]-Glu-Phe-Ile-Ala-Trp-Leu-Val-Arg-Gly-Arg-Gly-OH , Application: Glucagon-Like Peptide-1 (GLP-1) Analog Shown to Stimulate Insulin and Suppress Glucagon Secretion in a Glucose-Dependent Manner, Salt Form:, Acetate Salt, Storage Temperature: -20 °C, References: S.P. Marso, et al., N. Engl. J. Med., 375, 1834 (2016). , J. Lau, et al., J. Med. Chem., 58, 7370 (2015)., C.F. Gotfredsen, et al., Diabetes, 63, 2486: (2014).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.