Learn More
Microsolv Technology Corporation Semi Prep HPLC Column, HPS Amino, 5um, 120A. 10mm ID x 50mm Length. 1 EA.
Supplier:  Microsolv Technology Corporation 75040SP50
Encompass_Preferred
This Cogent HPS Amino™ Stationary Phase has been a premium value and used by HPLC laboratories for many years. Not end capped, it is manufactured using tri-functional bonding of a 3 carbon silane with a terminal amino group to our unique High Purity Silica. This Column is ideal for new, small volume prep methods. With a 5 µm Particle, this Column works well with higher flow rates and lower back pressures compared with smaller particles which is desirable for Semi Preparative Chromatography. Used for polar molecules with a negative charge, such compounds with Carboxylic Acid Groups and Sugars, in Normal Phase or HILIC modes. Compares to Inertsil™ Amino and Luna™ Amino Columns in performance. A specialized HPLC System is required for columns this size.
Catalog No. 50-227-1052
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.