Learn More
Brady™ Semiconductor and Chemical Pipe Marker: CITY WATER
Catalog No. 19112363 Shop All Brady Corporation Products
Change view
Click to view available options
Display Text:
CITY WATER
Catalog No. Display Text
19-112-363 CITY WATER
1 options
Catalog No. 19-112-363 Supplier Brady™ Supplier No. 91913
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Identify pipe content and flow direction. Brady™ Semiconductor and Chemical Pipe Markers are perfect for use on small diameter piping or tubing.
  • Meets ASME A13.1 color specifications
  • Made from B-946 self-sticking vinyl with permanent adhesive
  • Use with 1 in. arrow tape
Specifications
Description Abrasion and chemical resistant material
Adhesive Type Acrylic, Permanent, Cold-temperature
Color White, Green
Format Die-cut
Material Pressure-sensitive Vinyl (B-946)
Product Type Semiconductor and Chemical Pipe Marker
Display Text CITY WATER
Quantity 28/Pk.
Size (English) 2.25 x 0.5 in. (W x H)
Thickness (English) 0.004 in.