Learn More
Description
- Made of polyvinyl chloride, eliminating latex protein without sacrificing strength, feel or function
- Beaded -cuff, latex-free for optimal barrier protection
- Elasticity provides maximum tactile sensitivity
Specifications
Specifications
| Certifications/Compliance | FDA 21 CFR Part 177, ASTM D5250 |
| Certifications/Compliance (Europe) | CE Class I (MD Regulation (EU) 2017/745), CE Category I (minimal risk, PPE Regulation (EU) 2016/425), EN 455 1-4, EN ISO 21420, EN ISO 374 1-5, EN 421 |
| Chemotherapeutic Rated | No |
| Applications | Healthcare |
| AQL | 1.5 |
| Color | White |
| Disposable | Single-use |
| Exterior Finish | No Texture |
| Finger Thickness (English) | 4.4 mil |
| Finger Thickness (Metric) | 0.11 mm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.