Learn More
Invitrogen™ SENP1 Polyclonal Antibody

Description
Suggested positive control: HeLa or NIH-3T3 whole cell lysate.
Specifications
Specifications
| Antigen | SENP1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | SENP1 |
| Gene Accession No. | P59110, Q9P0U3 |
| Gene Alias | 2310046A20Rik; D15Ertd528e; E330036L07Rik; SEN1; Senp1; sentrin/SUMO-specific protease SENP1; Sentrin-specific protease 1; SUMO-1 protease 2; SUMO1/sentrin specific peptidase 1; SUMO1/sentrin specific protease 1; Sumo1/sentrin/SMT3 specific peptidase 1; Supr2; suPr-2 |
| Gene Symbols | SENP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.