Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SENP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531055
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531055 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531055 Supplier Invitrogen™ Supplier No. PA531055
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Raji, K562, THP-1. Predicted reactivity: Mouse (100%), Rat (99%), Xenopus laevis (87%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The reversible posttranslational modification of proteins by the addition of small ubiquitin-like SUMO proteins is required for numerous biologic processes. SUMO-specific proteases, such as SENP3, are responsible for the initial processing of SUMO precursors to generate a C-terminal diglycine motif required for the conjugation reaction. They also have isopeptidase activity for the removal of SUMO from high molecular mass SUMO conjugates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SENP3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene Senp3
Gene Accession No. Q9H4L4
Gene Alias AA408656; DKFZP586K0919; DKFZp762A152; SENP3; sentrin/SUMO-specific protease 3; sentrin/SUMO-specific protease SENP3; Sentrin-specific protease 3; Smt3ip; Smt3ip1; Smt3-specific isopeptidase 1; SSP3; SUMO specific peptidase 3; SUMO/sentrin specific peptidase 3; SUMO/sentrin specific protease 3; SUMO1/sentrin/SMT3 specific peptidase 3; SUMO1/sentrin/SMT3 specific protease 3; SUMO-1-specific protease 3; SUSP3; Ulp1
Gene Symbols Senp3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human SENP3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26168
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.