Learn More
Invitrogen™ SENP6 Polyclonal Antibody

Description
Antibody detects endogenous levels of total SENP6.
Specifications
Specifications
| Antigen | SENP6 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | SENP6 |
| Gene Accession No. | Q6P7W0, Q9GZR1 |
| Gene Alias | 2810017C20Rik; E130319N12Rik; FKSG6; FLJ11355; FLJ11887; KIAA0389; KIAA0797; mKIAA0797; RP1-134M13.1; SENP6; sentrin/SUMO-specific protease SENP6; Sentrin-specific protease 6; Sentrin-specific protease 6-like protein; SSP1; SUMO specific peptidase 6; SUMO/sentrin specific peptidase 6; SUMO/sentrin specific protease 6; SUMO1/sentrin specific peptidase 6; SUMO1/sentrin specific protease 6; Sumo1/sentrin/SMT3 specific peptidase 6; SUMO-1-specific protease 1; Susp1 |
| Gene Symbols | SENP6 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.