Learn More
Sensidyne™ Accessories for Sampling Media
SDP
Catalog No. 19001516 Shop All Sensidyne Inc. Products
Encompass
Change view
Click to view available options
Product Type:
Filter Cassette Holder
Tube Holder Deluxe Kit
Tube Holder Starter Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-001-516 Tube Holder Deluxe Kit
19-001-517 Tube Holder Starter Kit
19-031427 Filter Cassette Holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-001-516 Supplier Sensidyne™ Supplier No. 800232
Add to Cart
Add to Cart
Sampling media accessories include filter cassette holder that attaches to a worker's collar to sample breathing zone.
Tube holder starter kit includes single-variable manifold, holders, ends, and tubing. Tube holder deluxe kit includes single-, dual-, and triple-variable manifolds; holders; ends; and tubing.
Specifications
For Use With (Equipment) Standard Activated Charcoal Tube
Product Type Tube Holder Deluxe Kit
Includes Four of 6 x 70mm, three each of 7 to 10 x 110mm, 7 to 10 x 150mm and 7 to 10 x 175mm, two each of 5 x 150mm and 7 x 130mm