Learn More
Sensidyne™ Detector Tubes: Chloropicrin
SDP
Catalog No. 19822884 Shop All Sensidyne Inc. Products
Click to view available options
Measurement Range:
0.05 to 16
Catalog No. Measurement Range
19-822-884 0.05 to 16
1 options
Catalog No. 19-822-884 Supplier Sensidyne™ Supplier No. 172S
Detector tubes
All detector tubes are sold in packs of 10.
Specifications
Components Measured Chloropicrin
Measurement Range 0.05 to 16
No. of Strokes 1 to 2
Quantity 10
Response Time 1.5 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C