Learn More
Sensidyne™ Detector Tubes: Butyl Acrylate
SDP
Catalog No. 19046154 Shop All Sensidyne Inc. Products
Click to view available options
Measurement Range:
2 to 60
Catalog No. Measurement Range
19-046-154 2 to 60
1 options
Catalog No. 19-046-154 Supplier Sensidyne™ Supplier No. 211U
Detector tubes
All detector tubes are sold in packs of 10.
Specifications
Components Measured Butyl Acrylate
Measurement Range 2 to 60
No. of Strokes 2
Quantity 10
Response Time 3 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C