Learn More
Sensidyne™ Detector Tubes: Butyl Acetate
SDP
Catalog No. 18067Q Shop All Sensidyne Inc. Products
Click to view available options
Measurement Range:
10 to 400
Catalog No. Measurement Range
18-067Q 10 to 400
1 options
This item is not returnable. View return policy
Catalog No. 18-067Q Supplier Sensidyne™ Supplier No. 138U
This item is not returnable. View return policy
Detector tubes
All detector tubes are sold in packs of 10.
Specifications
Components Measured Butyl Acetate
Measurement Range 10 to 400
No. of Strokes 1
Quantity 10
Response Time 1.5 min.
Temperature Range (English) 50°F to 104°F
Temperature Range (Metric) 10° to 40°C