Learn More
Sensidyne™ Detector Tubes: Formaldehyde
SDP
Catalog No. 19120685 Shop All Sensidyne Inc. Products
Click to view available options
Measurement Range:
0.1 to 4
Catalog No. Measurement Range
19-120-685 0.1 to 4
1 options
This item is not returnable. View return policy
Catalog No. 19-120-685 Supplier Sensidyne™ Supplier No. 171SC
This item is not returnable. View return policy
Detector tubes
All detector tubes are sold in packs of 10.
Specifications
Components Measured Formaldehyde
Measurement Range 0.1 to 4
No. of Strokes 5
Quantity 10
Response Time 5 min.
Temperature Range (English) 50°F to 104°F
Temperature Range (Metric) 10° to 40°C