Learn More
Sensidyne™ Detector Tubes: Acetaldehyde
SDP
Catalog No. 19818970 Shop All Sensidyne Inc. Products
Click to view available options
Measurement Range:
5 to 140
Catalog No. Measurement Range
19-818-970 5 to 140
1 options
Catalog No. 19-818-970 Supplier Sensidyne™ Supplier No. 133SB
Detector tubes
All detector tubes are sold in packs of 10.
Specifications
Components Measured Acetaldehyde
Measurement Range 5 to 140
No. of Strokes 1
Quantity 10
Response Time 1 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C