Learn More
Sensidyne™ Detector Tubes: Dimethyl, Ethyl and Methyl Amine
SDP
Catalog No. 19818998 Shop All Sensidyne Inc. Products
Click to view available options
Measurement Range:
1 to 20
Catalog No. Measurement Range
19-818-998 1 to 20
1 options
Catalog No. 19-818-998 Supplier Sensidyne™ Supplier No. 227S
Detector tubes
All detector tubes are sold in packs of 10.
Specifications
Components Measured Amines
Measurement Range 1 to 20
No. of Strokes 1
Quantity 10
Response Time 1 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C