Learn More
Sensidyne™ Detector Tubes: Mercury Vapor
SDP
Catalog No. 18071G Shop All Sensidyne Inc. Products
Click to view available options
Measurement Range:
0.1 to 10mg/m3
Catalog No. Measurement Range
18-071G 0.1 to 10mg/m3
1 options
Catalog No. 18-071G Supplier Sensidyne™ Supplier No. 142S
Detector tubes are sold in packs of 10
Specifications
Components Measured Mercury Vapor
Measurement Range 0.1 to 10mg/m3
No. of Strokes 1/5
Quantity 10
Response Time 5 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C